siRNA Details
Gene Symbol: Hdac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200045.1 | NP_001186974.1 | 7 | 932 | ||
XM_008764610.1 | XP_008762832.1 | 6 | 993 | |||
XM_008764611.1 | XP_008762833.1 | 7 | 3154 | |||
XM_008764616.1 | XP_008762838.1 | 6 | 978 | |||
GenBank mRNA | AB558551.1 | - | 7 | 932 | ||
FQ225774.1 | - | 3 | 213 |
