siRNA Details
Gene Symbol: Zfpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242627.1 | NP_001229556.1 | 10 | 1341 | ||
XM_008772646.1 | XP_008770868.1 | 9 | 4821 | |||
XM_008772647.1 | XP_008770869.1 | 9 | 4809 | |||
XM_008772649.1 | XP_008770871.1 | 8 | 1295 |
