siRNA Details
Gene Symbol: Anks4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127650.1 | NP_001121122.1 | 1 | 626 | ||
GenBank mRNA | BC166493.1 | - | 1 | 626 | 7110362 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Anks4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127650.1 | NP_001121122.1 | 1 | 626 | ||
GenBank mRNA | BC166493.1 | - | 1 | 626 | 7110362 |