siRNA Details
Gene Symbol: RGD1566052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115047.2 | NP_001108519.1 | 3 | 341 | ||
XM_008762071.1 | XP_008760293.1 | 3 | 381 | |||
GenBank mRNA | BC158667.1 | - | 3 | 341 | 8370237 |
Gene Symbol: LOC365839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749325.3 | XP_003749373.1 | 1 | 379 |
