siRNA Details
Gene Symbol: Stambpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115048.1 | NP_001108520.1 | 3 | 355 | ||
XM_006231296.2 | XP_006231358.1 | 3 | 402 | |||
XM_006231297.2 | XP_006231359.1 | 3 | 167 | |||
XM_006231299.2 | XP_006231361.1 | 3 | 539 | |||
XM_008760346.1 | XP_008758568.1 | 2 | 124 | |||
XM_008760347.1 | XP_008758569.1 | 1 | 380 | |||
XM_008760349.1 | XP_008758571.1 | 2 | 155 | |||
GenBank mRNA | BC158624.1 | - | 3 | 355 | 7454746 |

More Information
Other Species Targeted

