siRNA Details
Gene Symbol: Stambpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115048.1 | NP_001108520.1 | 6 | 953 | ||
XM_006231296.2 | XP_006231358.1 | 6 | 1000 | |||
XM_006231297.2 | XP_006231359.1 | 6 | 765 | |||
XM_006231299.2 | XP_006231361.1 | 6 | 1137 | |||
XM_008760346.1 | XP_008758568.1 | 5 | 722 | |||
XM_008760347.1 | XP_008758569.1 | 4 | 978 | |||
XM_008760349.1 | XP_008758571.1 | 5 | 753 | |||
GenBank mRNA | BC158624.1 | - | 6 | 953 | 7454746 |
