siRNA Details
Gene Symbol: Traf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271240.1 | NP_001258169.1 | 4 | 401 | ||
XM_006233999.2 | XP_006234061.1 | 7 | 758 | |||
XM_006234000.2 | XP_006234062.1 | 7 | 673 | |||
GenBank mRNA | BC158696.1 | - | 7 | 545 |
