siRNA Details
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762189.1 | XP_008760411.1 | 1 | 313 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762189.1 | XP_008760411.1 | 1 | 313 |