siRNA Details
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100158.2 | NP_001093628.1 | 9, 10 | 1051 | ||
GenBank mRNA | BC105875.1 | - | 9, 10 | 996 | 7130292 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100158.2 | NP_001093628.1 | 9, 10 | 1051 | ||
GenBank mRNA | BC105875.1 | - | 9, 10 | 996 | 7130292 |