siRNA Details
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100158.2 | NP_001093628.1 | 4, 5 | 432 | ||
GenBank mRNA | BC105875.1 | - | 4, 5 | 377 | 7130292 | |
CO559861.1 | - | 4, 5 | 432 | |||
FQ234161.1 | - | 3, 4 | 153 |
