siRNA Details
Gene Symbol: Ccdc137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143896.1 | NP_001137368.1 | 5 | 651 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143896.1 | NP_001137368.1 | 5 | 651 |