siRNA Details
Gene Symbol: LOC688335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256150.1 | NP_001243079.1 | 28 | |||
GenBank mRNA | HQ840741.1 | - | 28 |
Gene Symbol: LOC100360143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729890.4 | XP_002729936.2 | 1323 |
Gene Symbol: LOC100911945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766076.1 | XP_008764298.1 | 1 | 1323 |
