siRNA Details
Gene Symbol: LOC100911945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766076.1 | XP_008764298.1 | 2 | 1373 |
Gene Symbol: LOC100360143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729890.4 | XP_002729936.2 | 1373 |
Gene Symbol: LOC100362690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256149.1 | NP_001243078.1 | 2 | 78 | ||
GenBank mRNA | HQ840742.1 | - | 2 | 78 |
Gene Symbol: LOC688335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256150.1 | NP_001243079.1 | 78 | |||
GenBank mRNA | HQ840741.1 | - | 78 |
