siRNA Details
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127572.1 | NP_001121044.1 | 14 | 1081 | ||
GenBank mRNA | BC161949.1 | - | 14 | 1081 | 9025530 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127572.1 | NP_001121044.1 | 14 | 1081 | ||
GenBank mRNA | BC161949.1 | - | 14 | 1081 | 9025530 |