siRNA Details
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127638.1 | NP_001121110.1 | 12 | 1783 | ||
XM_008769764.1 | XP_008767986.1 | 12 | 1926 | |||
GenBank mRNA | BC166443.1 | - | 12 | 1783 | 8363035 |
