siRNA Details
Gene Symbol: Kdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239015.2 | XP_006239077.1 | 1 | 653 | ||
XM_006239017.2 | XP_006239079.1 | 1 | 653 |
Gene Symbol: LOC100911383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773878.1 | XP_008772100.1 | 653 | |||
XM_008773880.1 | XP_008772102.1 | 653 |
