siRNA Details
Gene Symbol: Rbm24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191100.1 | NP_001178029.1 | 2 | 334 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbm24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191100.1 | NP_001178029.1 | 2 | 334 |