siRNA Details
Gene Symbol: Dip2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191564.1 | NP_001178493.1 | 2 | 181 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dip2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191564.1 | NP_001178493.1 | 2 | 181 |