siRNA Details
Gene Symbol: Stk38l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083336.1 | NP_001076805.1 | 8, 9 | 819 | ||
XM_008763431.1 | XP_008761653.1 | 7, 8 | 1716 | |||
GenBank mRNA | BC160812.1 | - | 6, 7 | 701 | 7460150 | |
EF444939.1 | - | 8, 9 | 819 |
