siRNA Details
Gene Symbol: Maz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110319.1 | NP_001103789.1 | 2 | 870 | ||
GenBank mRNA | EF059745.1 | - | 2 | 870 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010772, DQ150586, L04649, BC004758.
Gene Ontology Categories:
