siRNA Details
Gene Symbol: Mlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766417.1 | XP_008764639.1 | 12 | 3040 | ||
XM_008766418.1 | XP_008764640.1 | 11 | 2884 | |||
XM_008766420.1 | XP_008764642.1 | 12 | 2848 |
