siRNA Details
Gene Symbol: Sqrdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047913.2 | NP_001041378.2 | 4, 5 | 683 | ||
XM_006234887.2 | XP_006234949.1 | 5, 6 | 755 | |||
GenBank mRNA | BC107459.1 | - | 5, 6 | 691 | ||
BC158559.1 | - | 4, 5 | 683 | 7456444 |
