siRNA Details
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767925.1 | XP_008766147.1 | 11 | 3071 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767925.1 | XP_008766147.1 | 11 | 3071 |