siRNA Details
Gene Symbol: Gpr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001071777.1 | NP_001065245.1 | 1 | 333 | ||
XM_006254604.2 | XP_006254666.1 | 2 | 1850 | |||
GenBank mRNA | DQ777767.1 | - | 1 | 333 |
