siRNA Details
Gene Symbol: Car14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109655.2 | NP_001103125.1 | 10 | 913 | ||
XM_006232988.2 | XP_006233050.1 | 11 | 1458 | |||
XM_006232989.2 | XP_006233051.1 | 11 | 1436 | |||
XM_006232990.2 | XP_006233052.1 | 11 | 1418 | |||
GenBank mRNA | EF187253.1 | - | 10 | 913 |
