siRNA Details
Gene Symbol: Trim23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760683.1 | XP_008758905.1 | 5 | 625 | ||
XM_008760684.1 | XP_008758906.1 | 5 | 625 | |||
GenBank mRNA | FM052834.1 | - | 4 | 528 |
