siRNA Details
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031012.1 | NP_112274.1 | 16 | 2302 | ||
XM_006229407.2 | XP_006229469.1 | 17 | 2722 | |||
XM_006229408.2 | XP_006229470.1 | 17 | 2743 | |||
XM_006229409.1 | XP_006229471.1 | 17 | 2779 | |||
GenBank mRNA | M25073.1 | - | 16 | 2302 | ||
M26710.1 | - | 16 | 2349 |
