siRNA Details
Gene Symbol: Nell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031070.2 | NP_112332.2 | 12 | 1228 | ||
XM_008765797.1 | XP_008764019.1 | 11 | 1578 | |||
XM_008765798.1 | XP_008764020.1 | 12 | 1199 | |||
GenBank mRNA | AY089719.1 | - | 12 | 1228 | ||
BC093617.1 | - | 11 | 1172 | 7312010 | ||
EF110908.1 | - | 10 | 1021 | |||
U48245.1 | - | 12 | 1228 |
