siRNA Details
Gene Symbol: S100a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031114.1 | NP_112376.1 | 3 | 287 | ||
GenBank mRNA | AB453238.1 | - | 2 | 242 | ||
BC091565.1 | - | 3 | 313 | 7441806 | ||
FQ222734.1 | - | 3 | 342 | |||
FQ222778.1 | - | 3 | 310 | |||
FQ223502.1 | - | 3 | 307 | |||
FQ228469.1 | - | 3 | 310 | |||
FQ228745.1 | - | 3 | 310 | |||
FQ229216.1 | - | 3 | 310 | |||
J03627.1 | - | 3 | 287 |
