siRNA Details
Gene Symbol: Ocln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031329.2 | NP_112619.2 | 2, 3 | 199 | ||
XM_006231853.2 | XP_006231915.1 | 2, 3 | 195 | |||
XM_006231854.2 | XP_006231916.1 | 2, 3 | 239 | |||
XM_006231855.2 | XP_006231917.1 | 1, 2 | 119 | |||
GenBank mRNA | BC062037.1 | - | 2, 3 | 199 | 5623744 |
