siRNA Details
Gene Symbol: Rrad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053338.2 | NP_445790.2 | 2 | 390 | ||
XM_006255055.2 | XP_006255117.1 | 2 | 472 | |||
GenBank mRNA | CK602305.1 | - | 2 | 362 | ||
FM054321.1 | - | 2 | 443 | |||
U12187.1 | - | 2 | 362 |
