siRNA Details
Gene Symbol: Abcb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031760.1 | NP_113948.1 | 10 | 1070 | ||
GenBank mRNA | AF010597.1 | - | 10 | 1043 | ||
U69487.1 | - | 10 | 1070 |

More Information
Other Species Targeted

Gene Ontology Categories:
