siRNA Details
Gene Symbol: Gabbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031802.1 | NP_113990.1 | 7 | 1304 | ||
GenBank mRNA | AF058795.1 | - | 7 | 1304 | ||
AF074482.1 | - | 7 | 1154 | |||
AF109405.1 | - | 7 | 1271 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081141.
Set Membership: |
![]() |
Gene Ontology Categories:
