siRNA Details
Gene Symbol: Nr5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191099.1 | NP_001178028.1 | 4 | 993 | ||
XM_006234106.2 | XP_006234168.1 | 2 | 2004 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nr5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191099.1 | NP_001178028.1 | 4 | 993 | ||
XM_006234106.2 | XP_006234168.1 | 2 | 2004 |