siRNA Details
Gene Symbol: Klf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053394.2 | NP_445846.2 | 2 | 838 | ||
XM_006252403.2 | XP_006252465.1 | 2 | 475 | |||
GenBank mRNA | BC081758.1 | - | 2 | 480 | 7132572 |
