siRNA Details
Gene Symbol: Trpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083115.1 | NP_001076584.1 | 7, 8 | 2093 | ||
NM_080396.1 | NP_536321.1 | 7, 8 | 2093 | |||
XM_006232356.2 | XP_006232418.1 | 8, 9 | 2417 | |||
GenBank mRNA | AB008889.2 | - | 8, 9 | 2139 | ||
AF288407.1 | - | 7, 8 | 2109 | |||
AF288408.1 | - | 7, 8 | 2109 | |||
AF421363.1 | - | 7, 8 | 2093 | |||
AF421364.1 | - | 7, 8 | 2093 | |||
AF421365.1 | - | 7, 8 | 2093 | |||
AF421366.1 | - | 7, 8 | 2025 | |||
AF421367.1 | - | 7, 8 | 2040 | |||
AF421368.1 | - | 7, 8 | 2082 | |||
FQ228645.1 | - | 8, 9 | 2312 |
