siRNA Details
Gene Symbol: S100a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053485.2 | NP_445937.1 | 3 | 295 | ||
XM_006232779.2 | XP_006232841.1 | 3 | 390 | |||
GenBank mRNA | AF140232.1 | - | 3 | 295 | ||
AJ132717.1 | - | 2 | 268 | |||
BC166598.1 | - | 3 | 295 | 7931905 | ||
FQ221024.1 | - | 3 | 319 | |||
FQ221161.1 | - | 3 | 319 | |||
FQ221241.1 | - | 3 | 319 | |||
FQ221410.1 | - | 3 | 319 | |||
FQ221675.1 | - | 3 | 321 | |||
FQ221739.1 | - | 3 | 318 | |||
FQ221833.1 | - | 3 | 319 | |||
FQ221948.1 | - | 3 | 319 | |||
FQ222071.1 | - | 3 | 351 | |||
FQ222088.1 | - | 3 | 351 | |||
FQ222553.1 | - | 3 | 319 | |||
FQ222560.1 | - | 3 | 319 | |||
FQ223102.1 | - | 3 | 351 | |||
FQ223404.1 | - | 3 | 350 | |||
FQ223495.1 | - | 3 | 319 | |||
U31867.1 | - | 2 | 286 |

More Information
Other Species Targeted

Gene Ontology Categories:
