siRNA Details
Gene Symbol: Slc44a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033852.1 | NP_001029024.1 | 10 | 1245 | ||
NM_053492.3 | NP_445944.2 | 10 | 1245 | |||
XM_008763716.1 | XP_008761938.1 | 10 | 1278 | |||
XM_008763717.1 | XP_008761939.1 | 10 | 1279 | |||
GenBank mRNA | AJ245619.1 | - | 10 | 1237 | ||
AJ420809.1 | - | 10 | 1245 |
