siRNA Details
Gene Symbol: Slc25a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053515.1 | NP_445967.1 | 3 | 829 | ||
GenBank mRNA | BC060533.1 | - | 3 | 833 | 6922056 | |
D12770.1 | - | 3 | 796 | |||
FQ213027.1 | - | 3 | 826 | |||
FQ213612.1 | - | 3 | 855 | |||
FQ214384.1 | - | 3 | 825 | |||
FQ215448.1 | - | 3 | 828 | |||
FQ217254.1 | - | 3 | 825 | |||
FQ220525.1 | - | 3 | 824 | |||
FQ224814.1 | - | 3 | 826 |
