siRNA Details
Gene Symbol: Ngb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033359.3 | NP_203523.2 | 1 | 432 | ||
XM_008764864.1 | XP_008763086.1 | 1 | 1113 | |||
GenBank mRNA | AF333245.1 | - | 1 | 23 | ||
AF334379.1 | - | 1 | 23 | |||
AY066001.1 | - | 1 | 232 |
