siRNA Details
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053571.1 | NP_446023.1 | 15, 16 | 1989 | ||
XM_008769682.1 | XP_008767904.1 | 21, 22 | 5518 | |||
XM_008769683.1 | XP_008767905.1 | 19, 20 | 5226 | |||
XM_008769684.1 | XP_008767906.1 | 16, 17 | 2750 | |||
XM_008769685.1 | XP_008767907.1 | 18, 19 | 4503 | |||
XM_008769686.1 | XP_008767908.1 | 14, 15 | 1787 | |||
GenBank mRNA | AB060653.1 | - | 15, 16 | 1989 |
