siRNA Details
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053571.1 | NP_446023.1 | 18, 19 | 2322 | ||
XM_008769682.1 | XP_008767904.1 | 24, 25 | 5851 | |||
XM_008769683.1 | XP_008767905.1 | 22, 23 | 5559 | |||
XM_008769684.1 | XP_008767906.1 | 19, 20 | 3083 | |||
XM_008769685.1 | XP_008767907.1 | 21, 22 | 4836 | |||
XM_008769686.1 | XP_008767908.1 | 17, 18 | 2120 | |||
GenBank mRNA | AB060653.1 | - | 18, 19 | 2322 |
