siRNA Details
Gene Symbol: Mirg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_028265.1 | 2, 3 | 167 | |||
GenBank mRNA | AI605771.1 | - | 2, 3 | 167 | ||
AJ517767.1 | - | LIT1749 Mirg |
2, 3 | 167 | ||
EU616812.1 | - | 2751 | ||||
EU616813.1 | - | 2664 |
