siRNA Details
Gene Symbol: Tmem51os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027137.1 | 5 | 1580 | |||
GenBank mRNA | AK153738.1 | - | 2 | 3303 | ||
AK154034.1 | - | 5 | 1538 | |||
AK158244.1 | - | 4 | 1954 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY763106.1 | - | 2 | 578 | Chr. 4 - 142086892 - 142088045 [+] on Build GRCm38 |
