siRNA Details
Gene Symbol: AW011738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030671.1 | 1 | 1085 | |||
GenBank mRNA | AK036383.1 | - | 1 | 1066 | ||
AK078660.1 | - | 1 | 1066 | |||
AK085774.1 | - | 1066 | ||||
AK086145.1 | - | 1 | 1086 | |||
AK156747.1 | - | 1 | 1072 | |||
AK172062.1 | - | 2 | 901 | |||
AK172315.1 | - | 2 | 904 |
