siRNA Details
Gene Symbol: AW011738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030671.1 | 1 | 369 | |||
GenBank mRNA | AK036383.1 | - | 1 | 350 | ||
AK044300.1 | - | 351 | ||||
AK078660.1 | - | 1 | 351 | |||
AK083998.1 | - | 384 | ||||
AK085774.1 | - | 350 | ||||
AK086145.1 | - | 1 | 370 |
