siRNA Details
Gene Symbol: AW011738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030671.1 | 1 | 1743 | |||
GenBank mRNA | AK036383.1 | - | 1 | 1722 | ||
AK078660.1 | - | 1 | 1732 | |||
AK085774.1 | - | 1725 | ||||
AK172062.1 | - | 2 | 1563 | |||
AK172315.1 | - | 2 | 1632 |
