siRNA Details
Gene Symbol: AW011738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030671.1 | 1 | 2271 | |||
GenBank mRNA | AK078660.1 | - | 1 | 2280 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV356912.2 | - | 1 | 131 | Chr. 4 - 156205424 - 156206021 [+] on Build GRCm38 |
