siRNA Details
Gene Symbol: BC025920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030677.1 | 3 | 1418 | |||
GenBank mRNA | AK140772.1 | - | 3 | 1419 | ||
BC025920.1 | - | 2 | 2098 | |||
BC145724.1 | - | 2 | 1401 | |||
BC145726.1 | - | 2 | 1430 |
