siRNA Details
Gene Symbol: BC025920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030677.1 | 3 | 265 | |||
GenBank mRNA | AK140772.1 | - | 3 | 266 | ||
AK142150.1 | - | 1 | 1187 | |||
BC145724.1 | - | 2 | 248 | |||
BC145726.1 | - | 2 | 277 |
